siRNA Details
Gene Symbol: Lsm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015816.4 | NP_056631.2 | 5 | 897 | ||
GenBank mRNA | AJ249439.1 | - | 5 | 735 | ||
AK003847.1 | - | 5 | 733 | |||
AK010910.1 | - | 5 | 733 | |||
AK089028.1 | - | 5 | 750 | |||
AK143470.1 | - | 5 | 721 | |||
AK167784.1 | - | 5 | 761 | |||
BC026747.1 | - | 5 | 880 | 4024321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
