siRNA Details
Gene Symbol: Klf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021366.3 | NP_067341.2 | 2 | 1043 | ||
GenBank mRNA | AF251796.1 | - | 2 | 1042 | ||
AF252285.1 | - | 2 | 1021 | |||
AJ245644.1 | - | 2 | 978 | |||
AJ275987.1 | - | 2 | 991 | |||
AK002926.1 | - | 2 | 296 | |||
AK155298.1 | - | 2 | 994 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109147.
Gene Ontology Categories:
