siRNA Details
Gene Symbol: Tomm40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109748.1 | NP_001103218.1 | 9 | 1582 | ||
NM_016871.2 | NP_058567.2 | 10 | 1484 | |||
GenBank mRNA | AF043249.1 | - | 9 | 1260 | ||
AF316403.1 | - | 9 | 1417 | |||
AK142575.1 | - | 9 | 1178 | |||
AK150019.1 | - | 9 | 1579 | |||
AK164570.1 | - | 10 | 1468 | |||
AK165173.1 | - | 10 | 1479 | |||
AK170991.1 | - | 10 | 1468 | |||
BC100452.1 | - | 10 | 1472 | 6532792 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
