siRNA Details
Gene Symbol: Tsnax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016909.2 | NP_058605.1 | 5 | 660 | ||
GenBank mRNA | AF187040.1 | - | 5 | 660 | ||
AK014437.1 | - | 5 | 607 | |||
AK030407.1 | - | 5 | 590 | |||
AK140475.1 | - | 1 | 1490 | |||
AK146339.1 | - | 5 | 593 | |||
AK159223.1 | - | 5 | 662 | |||
AK164453.1 | - | 4 | 435 | |||
BC004611.1 | - | 5 | 562 | 3585400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
