siRNA Details
Gene Symbol: Rcan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286653.1 | NP_001273582.1 | 5 | 714 | ||
NM_001286654.1 | NP_001273583.1 | 5 | 591 | |||
NM_030598.2 | NP_085101.1 | 4 | 721 | |||
NM_207649.1 | NP_997532.1 | 5 | 847 | |||
GenBank mRNA | AB061524.1 | - | 4 | 659 | ||
AB061525.1 | - | 5 | 714 | |||
AF237791.1 | - | 4 | 461 | |||
AF237887.1 | - | 4 | 507 | |||
AK049006.1 | - | 4 | 660 | |||
AK136805.1 | - | 5 | 656 | |||
AK138768.1 | - | 5 | 759 | |||
AK160865.1 | - | 4 | 609 | |||
BC047153.1 | - | 4 | 519 | 6491318 | ||
BC049096.1 | - | 4 | 721 | 6416901 | ||
BC062141.1 | - | 5 | 591 | 30290791 | ||
BF534085.1 | - | 696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
