siRNA Details
Gene Symbol: Cpsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291248.1 | NP_001278177.1 | 8 | 1518 | ||
NM_001291249.1 | NP_001278178.1 | 8 | 1515 | |||
NM_178576.3 | NP_848671.1 | 7 | 1419 | |||
GenBank mRNA | AF033201.1 | - | 8 | 1219 | ||
AK046064.1 | - | 7 | 1409 | |||
BC025045.1 | - | 1 | 2486 | |||
BC057067.1 | - | 6 | 2773 | 5689404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
