siRNA Details
Gene Symbol: Rabep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291141.1 | NP_001278070.1 | 14, 15 | 2362 | ||
NM_001291142.1 | NP_001278071.1 | 14, 15 | 2371 | |||
NM_001291143.1 | NP_001278072.1 | 16, 17 | 2562 | |||
NM_019400.3 | NP_062273.2 | 15, 16 | 2491 | |||
GenBank mRNA | AB001750.1 | - | 16, 17 | 2398 | ||
AF248489.1 | - | 14, 15 | 2157 | |||
AF248490.1 | - | 14, 15 | 2148 | |||
AK134587.1 | - | 15, 16 | 2492 | |||
BC003921.1 | - | 6, 7 | 1112 | |||
BC060166.1 | - | 14, 15 | 3041 | |||
BC080274.1 | - | 15, 16 | 2463 | |||
BC129853.1 | - | 15, 16 | 2316 | 40109688 | ||
BC129854.1 | - | 15, 16 | 2310 | 40109689 | ||
D86066.1 | - | 15, 16 | 2327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
