siRNA Details
Gene Symbol: Rabep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291141.1 | NP_001278070.1 | 3 | 549 | ||
NM_001291142.1 | NP_001278071.1 | 4 | 678 | |||
NM_001291143.1 | NP_001278072.1 | 5 | 749 | |||
NM_019400.3 | NP_062273.2 | 4 | 678 | |||
GenBank mRNA | AB001750.1 | - | 5 | 585 | ||
AF248489.1 | - | 4 | 464 | |||
AF248490.1 | - | 3 | 335 | |||
AK019413.1 | - | 4 | 652 | |||
AK134587.1 | - | 4 | 677 | |||
AK137236.1 | - | 4 | 643 | |||
AK151901.1 | - | 4 | 647 | |||
BC009814.1 | - | 4 | 674 | |||
BC060166.1 | - | 4 | 657 | |||
BC080274.1 | - | 4 | 651 | |||
BC129853.1 | - | 4 | 497 | 40109688 | ||
BC129854.1 | - | 4 | 497 | 40109689 | ||
D86066.1 | - | 4 | 514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
