siRNA Details
Gene Symbol: Rpl36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018730.3 | NP_061200.2 | 3 | 234 | ||
GenBank mRNA | AK003115.1 | - | 3 | 214 | ||
BC021595.1 | - | 3 | 202 | 5100307 | ||
BC086914.1 | - | 3 | 213 | 6768133 | ||
CF728650.1 | - | 2 | 311 | |||
X75895.1 | - | 2 | 198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
