siRNA Details
Gene Symbol: Serpinb9f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183197.1 | NP_899020.1 | 7 | 1071 | ||
GenBank mRNA | BC052216.1 | - | 7 | 1071 | 30020424 | |
U96708.1 | - | 1 | 30 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
