siRNA Details
Gene Symbol: Rps14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020600.4 | NP_065625.2 | 5 | 565 | ||
GenBank mRNA | AK003148.1 | - | 5 | 510 | ||
AK011213.1 | - | 5 | 510 | |||
AK012425.1 | - | 5 | 509 | |||
AK012584.1 | - | 5 | 509 | |||
AK019233.1 | - | 5 | 508 | |||
AK028106.1 | - | 5 | 513 | |||
AK146325.1 | - | 5 | 506 | |||
AK146976.1 | - | 5 | 509 | |||
AK152461.1 | - | 1 | 3868 | |||
AK153401.1 | - | 1 | 3868 | |||
AK168237.1 | - | 5 | 509 | |||
AV490910.1 | - | 3 | 213 | |||
BC042940.1 | - | 4 | 489 | 6487681 | ||
BC062874.1 | - | 4 | 488 | 5683900 | ||
BC081449.1 | - | 4 | 476 | 5684375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
