siRNA Details
Gene Symbol: Gm13043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039595.2 | NP_001034684.1 | 3 | 1403 | ||
GenBank mRNA | BC099534.1 | - | 3 | 1403 | 30462179 |
Gene Symbol: BC080695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007579.3 | NP_001007580.1 | 1450 | |||
GenBank mRNA | BC080695.1 | - | 1451 | 30458858 |
Gene Symbol: Gm13040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113736.1 | NP_001107208.1 | 4 | 1450 |
Gene Symbol: Gm13057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113735.1 | NP_001107207.1 | 1450 |
Gene Symbol: Gm13119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034101.2 | NP_001029273.2 | 4 | 1421 | ||
GenBank mRNA | BC100406.1 | - | 3 | 1421 | 30727972 |
Gene Symbol: Pramef25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126315.2 | NP_001119787.1 | 3 | 1425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
