siRNA Details
Gene Symbol: Sod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011434.1 | NP_035564.1 | 5 | 524 | ||
GenBank mRNA | AK020624.1 | - | 5 | 430 | ||
AK077284.1 | - | 5 | 464 | |||
BC002066.1 | - | 5 | 478 | 3496016 | ||
BC048874.1 | - | 5 | 524 | 5401293 | ||
BC057074.1 | - | 3 | 3866 | |||
BC069974.1 | - | 3 | 586 | |||
BC086886.1 | - | 5 | 516 | 6760550 | ||
M35725.1 | - | 5 | 412 | |||
X06683.1 | - | 5 | 412 |

More Information
Other Species Targeted

Gene Ontology Categories:
