siRNA Details
Gene Symbol: Wdr45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290792.1 | NP_001277721.1 | 9 | 969 | ||
NM_001290794.1 | NP_001277723.1 | 9 | 865 | |||
NM_001290795.1 | NP_001277724.1 | 9 | 927 | |||
NM_172372.2 | NP_758960.1 | 9 | 965 | |||
GenBank mRNA | AF229635.1 | - | 9 | 901 | ||
AK008427.1 | - | 1 | 14 | |||
AK018409.1 | - | 9 | 964 | |||
AK145616.1 | - | 9 | 865 | |||
BC011479.1 | - | 9 | 936 | 4035468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
