siRNA Details
Gene Symbol: Otud5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290536.1 | NP_001277465.1 | 4 | 1133 | ||
NM_001290537.1 | NP_001277466.1 | 5 | 753 | |||
NM_138604.3 | NP_613070.2 | 4 | 1133 | |||
GenBank mRNA | AF229642.1 | - | 4 | 274 | ||
AK010750.1 | - | 5 | 751 | |||
AK149777.1 | - | 5 | 754 | |||
AK155131.1 | - | 4 | 1109 | |||
AK167226.1 | - | 3 | 446 | |||
AK171874.1 | - | 4 | 874 | |||
BC016529.1 | - | 4 | 1133 | 3594583 | ||
BC051111.1 | - | 4 | 872 | 5253679 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
