siRNA Details
Gene Symbol: Gm14484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025260.2 | NP_001020431.1 | 1 | 205 | ||
GenBank mRNA | BC049545.1 | - | 205 | 6770892 |
Gene Symbol: 1700012L04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029588.3 | NP_083864.1 | 1 | 243 | ||
GenBank mRNA | AK005922.1 | - | 1 | 245 |
Gene Symbol: Gm14474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242947.1 | NP_001229876.1 | 1 | 205 |
Gene Symbol: Gm14475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242954.1 | NP_001229883.1 | 148 |
Gene Symbol: Gm14476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242950.1 | NP_001229879.1 | 148 |
Gene Symbol: Gm14477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242949.1 | NP_001229878.1 | 148 |
Gene Symbol: Gm14478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242953.1 | NP_001229882.1 | 187 |
Gene Symbol: Gm14479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242951.1 | NP_001229880.1 | 148 |
Gene Symbol: Gm14482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242952.1 | NP_001229881.1 | 1 | 217 |
Gene Symbol: Gm14483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111037.1 | NP_001104507.1 | 1 | 212 | ||
GenBank mRNA | BC087913.1 | - | 1 | 212 | 6705184 |
Gene Symbol: Gm14501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085537.2 | NP_001079006.1 | 1 | 245 |
Gene Symbol: Gm5132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085517.2 | NP_001078986.1 | 1 | 191 |
Gene Symbol: Gm5382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034100.1 | NP_001029272.1 | 1 | 209 | ||
GenBank mRNA | BC100482.1 | - | 1 | 209 | 30918325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
