siRNA Details
Gene Symbol: LOC547349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025208.1 | NP_001020379.1 | 2 | 421 | ||
GenBank mRNA | AY533669.1 | - | 2 | 394 | ||
AY989869.1 | - | 2 | 421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
