siRNA Details
Gene Symbol: Cldn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165925.1 | NP_001159397.1 | 2 | 773 | ||
NM_001165926.1 | NP_001159398.1 | 2 | 963 | |||
NM_019500.4 | NP_062373.3 | 3 | 965 | |||
GenBank mRNA | AF124429.1 | - | 1 | 442 | ||
AF314089.2 | - | 3 | 965 | |||
AK020255.1 | - | 1 | 518 | |||
BC132183.1 | - | 3 | 883 | 40130460 | ||
BC138497.1 | - | 3 | 884 | 8861517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
