siRNA Details
Gene Symbol: Espn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019585.3 | NP_062531.2 | 5 | 848 | ||
NM_207687.2 | NP_997570.1 | 16 | 2586 | |||
NM_207688.2 | NP_997571.1 | 11 | 1615 | |||
NM_207689.2 | NP_997572.1 | 10 | 1372 | |||
NM_207690.2 | NP_997573.1 | 9 | 1345 | |||
NM_207691.2 | NP_997574.2 | 10 | 1588 | |||
GenBank mRNA | AF134857.1 | - | 4 | 321 | ||
AF239886.1 | - | 16 | 2586 | |||
AF540942.1 | - | 10 | 1588 | |||
AF540943.1 | - | 11 | 1615 | |||
AF540944.1 | - | 9 | 1345 | |||
AF540945.1 | - | 10 | 1372 | |||
AK002284.1 | - | 5 | 809 | |||
AK165010.1 | - | 7 | 2097 | |||
AY587570.1 | - | 7 | 865 | |||
AY587571.1 | - | 6 | 955 | |||
BC044723.1 | - | 5 | 848 | |||
BC057888.1 | - | 5 | 794 | |||
CJ194793.1 | - | 3 | 215 | |||
CJ319194.1 | - | 1 | 142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
