siRNA Details
Gene Symbol: Fam60a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019643.3 | NP_062617.2 | 3 | 300 | ||
GenBank mRNA | AK046519.1 | - | 3 | 207 | ||
AK145384.1 | - | 3 | 311 | |||
AK166569.1 | - | 3 | 306 | |||
BC076599.1 | - | 3 | 300 | 30614283 | ||
U64033.1 | - | 3 | 389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
