siRNA Details
Gene Symbol: Nupr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019738.1 | NP_062712.1 | 2 | 254 | ||
GenBank mRNA | AF131196.1 | - | 2 | 254 | ||
AK009572.1 | - | 2 | 274 | |||
BC002109.1 | - | 2 | 260 | 3487434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
