siRNA Details
Gene Symbol: Gltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019821.2 | NP_062795.2 | 2, 3 | 246 | ||
GenBank mRNA | AF209703.1 | - | 2, 3 | 172 | ||
AK157313.1 | - | 2, 3 | 233 | |||
BC016584.1 | - | 2, 3 | 246 | 3157800 | ||
BC053729.1 | - | 2, 3 | 189 | 30065192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
