siRNA Details
Gene Symbol: Apip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019735.4 | NP_062709.3 | 3 | 297 | ||
GenBank mRNA | AB028863.1 | - | 3 | 189 | ||
AK077705.1 | - | 3 | 298 | |||
AK145450.1 | - | 3 | 299 | |||
BC028434.1 | - | 3 | 240 | 1245515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
