siRNA Details
Gene Symbol: Dnaja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019794.4 | NP_062768.1 | 3 | 189 | ||
GenBank mRNA | AB028853.1 | - | 3 | 190 | ||
AK031858.1 | - | 3 | 256 | |||
AK077672.1 | - | 3 | 260 | |||
AK083208.1 | - | 3 | 257 | |||
AK163255.1 | - | 3 | 188 | |||
AK169169.1 | - | 3 | 249 | |||
BC003420.1 | - | 3 | 262 | 2582183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
