siRNA Details
Gene Symbol: Vapb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019806.5 | NP_062780.2 | 4, 5 | 523 | ||
GenBank mRNA | AF115504.1 | - | 4, 5 | 401 | ||
AK050632.1 | - | 4, 5 | 472 | |||
AK081908.1 | - | 4, 5 | 452 | |||
AK160925.1 | - | 4, 5 | 366 | |||
AK172080.1 | - | 4, 5 | 365 | |||
BC048231.1 | - | 4, 5 | 523 | 6313869 | ||
BC057123.1 | - | 4, 5 | 512 | 6411688 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_021847, XM_006235706, BC065576, AF086631.
Gene Ontology Categories:
