siRNA Details
Gene Symbol: Tspan6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019656.3 | NP_062630.2 | 5, 6 | 723 | ||
GenBank mRNA | AK146673.1 | - | 5, 6 | 628 | ||
AK149204.1 | - | 5, 6 | 633 | |||
AK160313.1 | - | 5, 6 | 617 | |||
AK167384.1 | - | 5, 6 | 619 | |||
BC003733.1 | - | 5, 6 | 657 | 3590862 | ||
BC110314.1 | - | 5, 6 | 632 | 5344878 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
