siRNA Details
Gene Symbol: Sec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271578.1 | NP_001258507.1 | 4 | 805 | ||
GenBank mRNA | AF113532.1 | - | 1 | 208 | ||
AK030246.1 | - | 4 | 808 | |||
BC100713.1 | - | 1 | 207 | |||
BC103557.1 | - | 1 | 207 | |||
BC103558.1 | - | 1 | 207 | |||
BC103559.1 | - | 1 | 207 | |||
BC115947.1 | - | 1 | 207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
