siRNA Details
Gene Symbol: Phax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162989.1 | NP_001156461.1 | 5 | 1042 | ||
NM_019996.4 | NP_064380.3 | 5 | 1152 | |||
GenBank mRNA | AJ276504.1 | - | 5 | 983 | ||
AK012949.1 | - | 5 | 1021 | |||
AK016958.1 | - | 5 | 1043 | |||
AK030749.1 | - | 5 | 1010 | |||
BC049590.1 | - | 5 | 998 | 6587693 | ||
BC061110.1 | - | 5 | 1012 | 30303763 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
