siRNA Details
Gene Symbol: Tob2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020507.3 | NP_065253.1 | 2 | 1211 | ||
GenBank mRNA | AK049766.1 | - | 2 | 1159 | ||
AK078476.1 | - | 1 | 52 | |||
AK143542.1 | - | 2 | 1188 | |||
BC011163.1 | - | 1 | 1093 | 4194136 | ||
BC094031.1 | - | 2 | 1142 | 4945656 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
