siRNA Details
Gene Symbol: DXBay18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025384.3 | NP_001020555.2 | 5 | 694 | ||
GenBank mRNA | AK005439.1 | - | 5 | 695 |
Gene Symbol: Gm14685 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025383.2 | NP_001020554.1 | 4 | 605 | ||
GenBank mRNA | BC094343.1 | - | 4 | 557 | 6402225 | |
BC147366.1 | - | 410 | 9056429 | |||
BC151162.1 | - | 410 | 9088051 |
