siRNA Details
Gene Symbol: Gmnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020567.2 | NP_065592.1 | 4 | 296 | ||
GenBank mRNA | AF068780.1 | - | 4 | 443 | ||
AK131959.1 | - | 4 | 270 | |||
BC020061.1 | - | 4 | 448 | 3988065 | ||
BY045588.1 | - | 4 | 297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
