siRNA Details
Gene Symbol: Stk32c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162540.1 | NP_001156012.1 | 7, 8 | 713 | ||
NM_021302.3 | NP_067277.2 | 7, 8 | 1013 | |||
GenBank mRNA | AB041542.1 | - | 7, 8 | 993 | ||
AK046439.1 | - | 7, 8 | 991 | |||
BC026457.1 | - | 7, 8 | 998 | 4481218 |
