siRNA Details
Gene Symbol: Ntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF268066.1 | - | 10 | 1790 | ||
AF281278.1 | - | 10 | 2043 | |||
BC119182.1 | - | 10 | 1877 | 8733931 | ||
BC119184.1 | - | 10 | 1877 | 8733933 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
