siRNA Details
Gene Symbol: Mrps30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021556.3 | NP_067531.1 | 4 | 931 | ||
GenBank mRNA | AK011476.1 | - | 4 | 933 | ||
AK013360.1 | - | 4 | 767 | |||
AK151642.1 | - | 4 | 932 | |||
AL355719.1 | - | 1 | 67 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
