siRNA Details
Gene Symbol: Zfp191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021559.2 | NP_067534.2 | 2 | 491 | ||
GenBank mRNA | AB221566.1 | - | 1 | 350 | ||
AF149093.1 | - | 1 | 350 | |||
AK146726.1 | - | 2 | 489 | |||
AY832930.1 | - | 2 | 530 | |||
BC011345.1 | - | 2 | 534 | 3992743 | ||
BC054832.1 | - | 2 | 491 | 6467818 |
Gene Symbol: Zf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK084191.1 | - | 1 | 89 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
