siRNA Details
Gene Symbol: Fxyd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022004.6 | NP_071287.1 | 5 | 499 | ||
GenBank mRNA | AA521976.1 | - | 5 | 238 | ||
AB032010.2 | - | 5 | 212 | |||
AK003888.1 | - | 5 | 262 | |||
AK018354.1 | - | 5 | 232 | |||
BC042579.1 | - | 5 | 245 | 3709336 | ||
BC051127.1 | - | 5 | 270 | 6531451 | ||
BQ891313.1 | - | 5 | 213 | |||
BY704145.1 | - | 5 | 256 | |||
W34202.1 | - | 5 | 219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
