siRNA Details
Gene Symbol: Nek6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159631.1 | NP_001153103.1 | 6 | 578 | ||
NM_021606.3 | NP_067619.1 | 6 | 571 | |||
GenBank mRNA | AF218847.1 | - | 6 | 573 | ||
AK004925.1 | - | 6 | 548 | |||
AK011519.1 | - | 6 | 557 | |||
AK029266.1 | - | 6 | 560 | |||
AK075717.1 | - | 6 | 545 | |||
AK075836.1 | - | 6 | 557 | |||
AK086700.1 | - | 576 | ||||
AK089919.1 | - | 6 | 532 | |||
AK170757.1 | - | 6 | 578 | |||
BC019524.1 | - | 6 | 640 | 4209097 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
