siRNA Details
Gene Symbol: Wtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113532.1 | NP_001107004.1 | 3, 4 | 273 | ||
NM_001113533.1 | NP_001107005.1 | 3, 4 | 273 | |||
NM_175394.2 | NP_780603.1 | 3, 4 | 321 | |||
GenBank mRNA | AJ276707.1 | - | 1, 2 | 31 | ||
AK031965.1 | - | 3, 4 | 329 | |||
AK076111.1 | - | 3, 4 | 315 | |||
AK156334.1 | - | 3, 4 | 169 | |||
BC046416.1 | - | 3, 4 | 202 | |||
BC047338.1 | - | 3, 4 | 202 | |||
BC093504.1 | - | 3, 4 | 206 | |||
BU516500.1 | - | 408 | ||||
BY016042.1 | - | 3, 4 | 273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
