siRNA Details
Gene Symbol: Zbed4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181412.3 | NP_852077.1 | 3 | 1173 | ||
GenBank mRNA | AK122335.1 | - | 3 | 1143 | ||
BC052174.1 | - | 2 | 1076 | 6518433 | ||
BC092524.1 | - | 1 | 797 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
