siRNA Details
Gene Symbol: Rps27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033865.1 | NP_001029037.1 | 5 | 777 | ||
NM_024277.2 | NP_077239.1 | 6 | 682 | |||
GenBank mRNA | AK018706.1 | - | 6 | 504 | ||
AK159742.1 | - | 1 | 141 | |||
BC002108.1 | - | 5 | 492 | 3487291 | ||
BC081446.1 | - | 5 | 483 | 6408051 | ||
BC096392.1 | - | 5 | 496 | 1380923 |

More Information
Other Species Targeted

Gene Ontology Categories:
