siRNA Details
Gene Symbol: N4BP2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320836.1 | NP_001307765.1 | 2 | 525 | ||
NM_014887.2 | NP_055702.1 | 2 | 525 | |||
XM_011534874.2 | XP_011533176.1 | 2 | 550 | |||
XM_017020324.1 | XP_016875813.1 | 3 | 491 | |||
XM_017020325.1 | XP_016875814.1 | 2 | 485 | |||
XM_017020326.1 | XP_016875815.1 | 2 | 370 | |||
XM_017020327.1 | XP_016875816.1 | 2 | 518 | |||
XM_017020328.1 | XP_016875817.1 | 3 | 491 | |||
XM_017020329.1 | XP_016875818.1 | 2 | 485 | |||
XM_017020330.1 | XP_016875819.1 | 2 | 518 | |||
XM_017020342.1 | XP_016875831.1 | 2 | 550 | |||
XM_017020347.1 | XP_016875836.1 | 3 | 491 | |||
XM_017020348.1 | XP_016875837.1 | 2 | 485 | |||
XM_017020349.1 | XP_016875838.1 | 2 | 370 | |||
GenBank mRNA | AF530063.1 | - | 1 | 347 | ||
AK312924.1 | - | 2 | 522 | |||
AL049783.1 | - | 2 | 512 | |||
BC010643.1 | - | 2 | 508 | 3852778 | ||
BG435390.1 | - | 2 | 526 | |||
BT007172.1 | - | 1 | 347 | |||
HY045074.1 | - | 2 | 456 | |||
U50532.1 | - | 2 | 515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
