siRNA Details
Gene Symbol: Gm6297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139286.1 | - | 1 | 1288 | ||
AK139295.1 | - | 1 | 822 | |||
AK172127.1 | - | 1 | 835 | |||
BE656271.1 | - | 2 | 298 | |||
CX215998.1 | - | 2 | 136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
