siRNA Details
Gene Symbol: Fcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028632.2 | NP_082908.2 | 7 | 518 | ||
GenBank mRNA | AK003561.1 | - | 6 | 477 | ||
BC094619.1 | - | 7 | 505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
