siRNA Details
Gene Symbol: Mgst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025569.1 | NP_079845.1 | 2 | 131 | ||
GenBank mRNA | AK003246.1 | - | 2 | 131 | ||
AK003309.1 | - | 2 | 117 | |||
AK003476.1 | - | 2 | 124 | |||
AK003492.1 | - | 2 | 124 | |||
AK008533.1 | - | 2 | 120 | |||
AK135742.1 | - | 2 | 106 | |||
BC029669.1 | - | 1 | 110 | 5372427 |

More Information
Other Species Targeted

Gene Ontology Categories:
