siRNA Details
Gene Symbol: Gm11487 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013393.1 | NP_001013411.1 | 5, 6 | 912 | ||
GenBank mRNA | CK332043.1 | - | 2, 3 | 378 |
Gene Symbol: Gm11757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085538.2 | NP_001079007.1 | 882 |
Gene Symbol: Gm11758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097978.2 | NP_001091447.2 | 914 | |||
GenBank mRNA | BC172045.1 | - | 5, 6 | 882 | 9054331 |
Gene Symbol: Gm13871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177578.2 | NP_001171049.1 | 5, 6 | 882 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
