siRNA Details
Gene Symbol: Atp5j2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020582.2 | NP_065607.1 | 2, 3 | 206 | ||
GenBank mRNA | AB030192.1 | - | 2, 3 | 207 | ||
AK002519.1 | - | 2, 3 | 156 | |||
AK003235.1 | - | 2, 3 | 144 | |||
AK003817.1 | - | 2, 3 | 156 | |||
AK013130.1 | - | 2, 3 | 156 | |||
AK151737.1 | - | 2, 3 | 155 | |||
AK168095.1 | - | 2, 3 | 156 | |||
BC029226.1 | - | 1, 2 | 146 | 2651401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
