siRNA Details
Gene Symbol: Gm14322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243903.1 | NP_001230832.1 | 5 | 2265 | ||
GenBank mRNA | BC092145.1 | - | 6 | 1359 |
Gene Symbol: Gm6182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA407120.1 | - | 199 | |||
AK008984.1 | - | 6 | 2050 | |||
AK163771.1 | - | 1 | 832 | |||
AK166086.1 | - | 5 | 1953 | |||
BC048160.1 | - | 5 | 1916 | |||
BC051922.1 | - | 5 | 1916 | |||
BC059210.1 | - | 4 | 3050 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
