siRNA Details
Gene Symbol: Gm6792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177416.1 | NP_001170887.1 | 14 | 508 | ||
NM_001177417.1 | NP_001170888.1 | 13 | 327 | |||
GenBank mRNA | AK079003.1 | - | 12 | 332 | ||
AV381130.2 | - | 3 | 213 | |||
AY226991.1 | - | 14 | 506 |
