siRNA Details
Gene Symbol: Anapc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291348.1 | NP_001278277.1 | 5 | 464 | ||
NM_001291349.1 | NP_001278278.1 | 4 | 414 | |||
NM_001291352.1 | NP_001278281.1 | 5 | 457 | |||
NM_001291353.1 | NP_001278282.1 | 5 | 457 | |||
NM_027532.4 | NP_081808.2 | 5 | 499 | |||
GenBank mRNA | AK014309.1 | - | 5 | 416 | ||
AK018169.1 | - | 4 | 404 | |||
AK135201.1 | - | 5 | 415 | |||
AK139635.1 | - | 5 | 456 | |||
BB636259.1 | - | 4 | 329 | |||
BC006736.1 | - | 5 | 457 | 3964006 | ||
BC080730.1 | - | 5 | 402 | 6507829 | ||
DV659149.1 | - | 5 | 469 |

More Information
Other Species Targeted

Gene Ontology Categories:
