siRNA Details
Gene Symbol: Igh-VX24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF099087.1 | - | 1 | 130 | ||
K00728.1 | - | 1 | 148 | |||
L41876.1 | - | 2 | 205 | |||
Z75771.1 | - | 1 | 125 | |||
Z75773.1 | - | 1 | 112 | |||
Z75779.1 | - | 1 | 125 | |||
Z75794.1 | - | 1 | 125 | |||
Z75796.1 | - | 1 | 125 | |||
Z75798.1 | - | 1 | 118 |
Gene Symbol: Ighg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156420.1 | - | 2 | 236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
